Learn More
Med Vet International Exel Needle, 16G X 5/8in, Hypodermic, Vet Use, 100/BX, 26455B
Catalog No. 502092509
Encompass_Preferred
This Exel International 26455B Needle is a 16G X 5/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2509 Supplier Med Vet International Supplier No. 26455B
This Exel International 26455B Needle is a 16G X 5/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX