Learn More
Med Vet International Nipro Hypodermic Needles, 18G x 1-in, 100/Box
Catalog No. 502092512
Encompass_Preferred
The Nipro AH+1825 18g x 1in. hypodermic needle has a thin wall that allows for rapid and efficient drug delivery. Nipro hypodermic needles are made with a proprietary grinding process that results in an exceptionally sharp lancet point that can penetrate tissue smoothly with minimal trauma. This needle has a pink colored hub for accurate, quick identification in or out of the package. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-209-2512 Supplier Med Vet International Supplier No. AH1825
The Nipro AH+1825 18g x 1in. hypodermic needle has a thin wall that allows for rapid and efficient drug delivery. Nipro hypodermic needles are made with a proprietary grinding process that results in an exceptionally sharp lancet point that can penetrate tissue smoothly with minimal trauma. This needle has a pink colored hub for accurate, quick identification in or out of the package. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.