Learn More
Med Vet International Exel Needle, 18G X 1.5in, Hypodermic, Vet Use, 100/BX, 26464
Catalog No. 502092518
Encompass_Preferred
This Exel International 26464 Needle is a 18G X 1.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2518 Supplier Med Vet International Supplier No. 26464
This Exel International 26464 Needle is a 18G X 1.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX