Learn More
Med Vet International Exel Needle, 18G X 3/4in, Hypodermic, Vet Use, 100/BX, 26461
Catalog No. 502092519
Encompass_Preferred
This Exel International 26461 Needle is a 18G X 3/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2519 Supplier Med Vet International Supplier No. 26461
This Exel International 26461 Needle is a 18G X 3/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX