Learn More
Med Vet International Terumo Needle, 19G x 1in, Hypodermic, 100/Box, NN1925R
Catalog No. 502092522
Encompass_Preferred
This Terumo NN1925R Needle comes with a 19G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2522 Supplier Med Vet International Supplier No. 1925RN
This Terumo NN1925R Needle comes with a 19G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX