Learn More
Med Vet International AHS Hypodermic Needles, 20G x 1.5in, 100/Box, AHS2038RN
Catalog No. 502092526
Encompass_Preferred
This AHS AHS2038RN Needle is a 20G X 1.5' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2526 Supplier Med Vet International Supplier No. AHS2038RN
This AHS AHS2038RN Needle is a 20G X 1.5' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX