Learn More
Med Vet International Terumo Needle, 20GX 1in, Hypodermic, 100/BX, NN2025R
Catalog No. 502092525
Encompass_Preferred
This Terumo NN2025R Needle comes with a 20GX 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX
Catalog No. 50-209-2525 Supplier Med Vet International Supplier No. 2025RN
This Terumo NN2025R Needle comes with a 20GX 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX