Learn More
Med Vet International Nipro Needle, 20G x 1in, Hypodermic, 100/Box, AH+2025
Catalog No. 502092527
Encompass_Preferred
This Nipro AH+2025 Needle is a 20G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2527 Supplier Med Vet International Supplier No. AH2025
This Nipro AH+2025 Needle is a 20G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX