Learn More
Med Vet International Exel Needle, 20G X 1in, Hypodermic, Vet Use, 100/BX, 26467
Supplier:  Med Vet International 26467
Encompass_Preferred
This Exel International 26467 Needle is a 20G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-255-9744