Learn More
Med Vet International Terumo Needle, 22GX 1 1/2in, Hypodermic, 100/BX, NN2238R
Catalog No. 502092543
Encompass_Preferred
This Terumo NN2238R Needle comes with a 22GX 1 1/2' Thin Wall needle. Terumo's thin wall needles areprecisely honed and beveled. This makes for ease of penetration and less patient discomfort This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2543 Supplier Med Vet International Supplier No. 2238RN
This Terumo NN2238R Needle comes with a 22GX 1 1/2' Thin Wall needle. Terumo's thin wall needles areprecisely honed and beveled. This makes for ease of penetration and less patient discomfort This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX