Learn More
Med Vet International AHS Hypodermic Needles, 22G x 1in, 100/Box, AHS2225RN
Catalog No. 502092544
Encompass_Preferred
This AHS AHS2225RN Needle is a 22 X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2544 Supplier Med Vet International Supplier No. AHS2225RN
This AHS AHS2225RN Needle is a 22 X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX