Learn More
Med Vet International Exel Needle, 22G X 1.5in, Multi-Sample Blood Draw, 100/Box, 26506
Catalog No. 502092555
Encompass_Preferred
This Exel International 26506 Needle is a 22G X 1.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This multi-sample needles permit several samples to be taken with a single puncture. They are also known as blood draw needles This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-2555 Supplier Med Vet International Supplier No. 26506
This Exel International 26506 Needle is a 22G X 1.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This multi-sample needles permit several samples to be taken with a single puncture. They are also known as blood draw needles This quantity is 100/BX