Learn More
Med Vet International Nipro Needle, 23G x 1in, Hypodermic, 100/Box, AH+2325
Catalog No. 502092558
Encompass_Preferred
This Nipro AH+2325 Needle is a 23G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2558 Supplier Med Vet International Supplier No. AH2325
This Nipro AH+2325 Needle is a 23G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX