Learn More
Med Vet International Terumo Needle, 25GX 5/8in, Hypodermic, 100/BX, NN2516R
Catalog No. 502092568
Encompass_Preferred
This Terumo NN2516R Needle comes with a 25GX 5/8' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX
Catalog No. 50-209-2568 Supplier Med Vet International Supplier No. 2516RN
This Terumo NN2516R Needle comes with a 25GX 5/8' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This quantity is 100/BX