Learn More
Med Vet International Terumo Needle, 25GX 1 1/2in, Hypodermic, 100/BX, NN2538R
Supplier:  Med Vet International 2538RN
Encompass_Preferred
This Terumo NN2538R Needle is a 25GX 1 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2571
 Restricted Item