Learn More
Med Vet International Exel Needle, 26G x 1/2in, Hypodermic, 100/Box, 26402
Catalog No. 502092575
Encompass_Preferred
This Exel International 26402 Needle is a 26G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-2575 Supplier Med Vet International Supplier No. 26402
 Restricted Item
Add to Cart
Add to Cart
This Exel International 26402 Needle is a 26G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX