Learn More
Med Vet International AHS Hypodermic Needles, 30G x 1/2 in, 100/Box, AHS3013RN
Catalog No. 502092583
Encompass_Preferred
This AHS AHS3013RN Needle is a 30G X 1/2" hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2583 Supplier Med Vet International Supplier No. AHS3013RN
This AHS AHS3013RN Needle is a 30G X 1/2" hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX