Learn More
Med Vet International Exel Needle, 30G x 1in, Hypodermic, 100/Box, 26439
Catalog No. 502092584
Encompass_Preferred
This Exel International 26439 Needle is a 30G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
This item is not returnable. View return policy
Catalog No. 50-209-2584 Supplier Med Vet International Supplier No. 26439
This Exel International 26439 Needle is a 30G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX