Learn More
Med Vet International Nipro Needle, 20G X 1in, Blood Collection, 100/Box, NM+20G25
Catalog No. 502092585
Encompass_Preferred
This Nipro NM+20G25 Needle is a 20G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. This is a multiple-draw blood collection needle. It is design to provide the convenience of several sample collections with a single vein puncture. This quantity is 100/BX
Catalog No. 50-209-2585 Supplier Med Vet International Supplier No. NM20G25
This Nipro NM+20G25 Needle is a 20G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. This is a multiple-draw blood collection needle. It is design to provide the convenience of several sample collections with a single vein puncture. This quantity is 100/BX