Learn More
Med Vet International Nipro Needle, 20G x 1-1/2in, Blood Collection, 100/Box, NM+20G38
Catalog No. 502092586
Encompass_Preferred
This Nipro NM+20G38 Needle is a 20G X 1-1/2' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. This is a multiple-draw blood collection needle. It is design to provide the convenience of several sample collections with a single vein puncture. This quantity is 100/Box
Catalog No. 50-209-2586 Supplier Med Vet International Supplier No. NM20G38
This Nipro NM+20G38 Needle is a 20G X 1-1/2' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. This is a multiple-draw blood collection needle. It is design to provide the convenience of several sample collections with a single vein puncture. This quantity is 100/Box