Learn More
Med Vet International Nipro Needle, 21G X 1in, Blood Collection, 100/Box, NM+21G25
Catalog No. 502092587
Encompass_Preferred
This Nipro NM+21G25 Needle is a 21G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. This is a multiple-draw blood collection needle. It is design to provide the convenience of several sample collections with a single vein puncture. This quantity is 100/Box
Catalog No. 50-209-2587 Supplier Med Vet International Supplier No. NM21G25
 Restricted Item
Add to Cart
Add to Cart
This Nipro NM+21G25 Needle is a 21G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. This is a multiple-draw blood collection needle. It is design to provide the convenience of several sample collections with a single vein puncture. This quantity is 100/Box