Learn More
Med Vet International Exel Needle, 30G X21MM, Dental, 100/BX, 26555
Supplier:  Med Vet International 26555
Encompass_Preferred
This Exel International 26555 Needle is a 30G X21MM Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a dental needle. It is used for intraoral injections. This quantity is 100/BX
Catalog No. 50-255-9735
 Restricted Item