Learn More
World Precision Instrument Dissecting Needle, 14 cm, Straight, Package of 12
SDP
Catalog No. 50822525
Encompass
A Dissecting Needle, also known as a dissecting probe or dissecting pin, is used in dissection procedures to manipulate and separate tissues, examine specimens, or perform precise dissection techniques. Our Dissecting Needle is 14 cm (5.5") long and has a plastic handle for a solid and comfortable grip. The thin, pointed metallic shaft is available as straight or angled. Dissecting needles are used to gently separate and lift tissues during dissection, or for holding or positioning small specimens or structures. With their fine and sharp tips, Dissecting Needles are also handy for microdissection procedures involving microscopic structures. The shaft is stainless steel, but these are not autoclavable. They are designed to be disposable. Each package contains 12 dissecting needles.
Catalog No. 50-822-525 Supplier World Precision Instrument Supplier No. 500458
A Dissecting Needle, also known as a dissecting probe or dissecting pin, is used in dissection procedures to manipulate and separate tissues, examine specimens, or perform precise dissection techniques. Our Dissecting Needle is 14 cm (5.5") long and has a plastic handle for a solid and comfortable grip. The thin, pointed metallic shaft is available as straight or angled. Dissecting needles are used to gently separate and lift tissues during dissection, or for holding or positioning small specimens or structures. With their fine and sharp tips, Dissecting Needles are also handy for microdissection procedures involving microscopic structures. The shaft is stainless steel, but these are not autoclavable. They are designed to be disposable. Each package contains 12 dissecting needles.