Learn More
Trajan Scientific NEEDLES LL MLL-5/23 5/PK
SDP
Catalog No. 502530258
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 51 mm, 2 in., Needle Gauge: 23 G, Needle Point Style: Bevel, Diameter Inner: 0.012 in., 0.32 mm, Diameter Outer: 0.024 in., 0.63 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0258 Supplier Trajan Scientific Supplier No. 039802
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 51 mm, 2 in., Needle Gauge: 23 G, Needle Point Style: Bevel, Diameter Inner: 0.012 in., 0.32 mm, Diameter Outer: 0.024 in., 0.63 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle