Learn More
Trajan Scientific NEEDLES N25/500 11.5/.063 5/PK
SDP
Catalog No. 502530261
Encompass_Preferred
Trajan Replacement Needles for 25-500uL Syringes, Length Needle: 115 mm, 4.52 in., Diameter Outer: 0.63 mm, 0.024 in., Diameter Inner: 0.24 mm, 0.009 in., Needle Point Style: Bevel, Needle Gauge: 23 G, For Use With: Chromatography, Stainless Steel, Quantity: 5/Pk.
Catalog No. 50-253-0261 Supplier Trajan Scientific Supplier No. 038161
Trajan Replacement Needles for 25-500uL Syringes, Length Needle: 115 mm, 4.52 in., Diameter Outer: 0.63 mm, 0.024 in., Diameter Inner: 0.24 mm, 0.009 in., Needle Point Style: Bevel, Needle Gauge: 23 G, For Use With: Chromatography, Stainless Steel, Quantity: 5/Pk.