Learn More
Trajan Scientific NEEDLES N5 5/0.63C
SDP
Catalog No. 502530266
Encompass_Preferred
Trajan Replacement Needles For 5ul Removable Needle Syringes, Length Needle: 50 mm, Needle Gauge: 23 G, Needle Point Style: Cone, Diameter Inner Needle: 0.11 mm, Diameter Outer Needle: 0.63 mm
Catalog No. 50-253-0266 Supplier Trajan Scientific Supplier No. 036011
Trajan Replacement Needles For 5ul Removable Needle Syringes, Length Needle: 50 mm, Needle Gauge: 23 G, Needle Point Style: Cone, Diameter Inner Needle: 0.11 mm, Diameter Outer Needle: 0.63 mm