Learn More
Trajan Scientific NEEDLES NLL 11.5/23 5/PK
SDP
Catalog No. 502530255
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 115 mm, 4.52 in., Needle Gauge: 23 G, Needle Point Style: Bevel, Diameter Inner: 0.012 in., 0.32 mm, Diameter Outer: 0.024 in., 0.63 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0255 Supplier Trajan Scientific Supplier No. 039811
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 115 mm, 4.52 in., Needle Gauge: 23 G, Needle Point Style: Bevel, Diameter Inner: 0.012 in., 0.32 mm, Diameter Outer: 0.024 in., 0.63 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle