Learn More
Med Vet International Dynarex Pen Needles, 30G, 5 mm, 1200/Case
Catalog No. 503043774
Encompass_Preferred
The Dynarex Pen Needles are individually packaged, sterile, and compatible with most insulin pens. The needles are available in a variety of gauges and lengths to accommodate a range of user needs. Ideal for use in a wide range of clinical and home healthcare environments. Dynarex Pen Needles are sterile, hollow needles specifically designed to easily deliver injectable medications. The lubricated head maximizes patient comfort, even with daily use.
Catalog No. 50-304-3774 Supplier Med Vet International Supplier No. 7024
The Dynarex Pen Needles are individually packaged, sterile, and compatible with most insulin pens. The needles are available in a variety of gauges and lengths to accommodate a range of user needs. Ideal for use in a wide range of clinical and home healthcare environments. Dynarex Pen Needles are sterile, hollow needles specifically designed to easily deliver injectable medications. The lubricated head maximizes patient comfort, even with daily use.