Learn More
Med Vet International AHS Hypodermic Needles, 18G x 1.5in, 100/Box, AHS1838RN
Catalog No. 502092598
Encompass_Preferred
This AHS AHS1838RN Needle is a 18G X 1.5" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2598 Supplier Med Vet International Supplier No. AHS1838RN
 Restricted Item
Add to Cart
Add to Cart
This AHS AHS1838RN Needle is a 18G X 1.5" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX