Learn More
Med Vet International EasyTouch Hypodermic Needles, 22G x 1in, 100/Box
Supplier:  Med Vet International 802201
Encompass_Preferred
Tri-bevel Cut Tip for Superior Sharpness Anti-coring Technology Electropolished to Remove Burrs Thin-walled for Maximum Flow Film-coated for a Comfortable Injection Fits All Standard Luer Lock & Slip Fit Syringe Barrels
Catalog No. 50-255-9681
 Restricted Item