Learn More
Med Vet International quincke Bevel spinal needles 25G x (0.5mm) x 3 1/2": Hub color Orange
Catalog No. 502092597
Encompass_Preferred
This Exel International 26970 Needle is a 25G X 3.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a spinal needle. It is used for entering the spinal canal or cisterna magna. This quantity is 1 each
This item is not returnable. View return policy
Catalog No. 50-209-2597 Supplier Med Vet International Supplier No. 26970
This Exel International 26970 Needle is a 25G X 3.5' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a spinal needle. It is used for entering the spinal canal or cisterna magna. This quantity is 1 each