Learn More
MSC NEEDLESTICK ARM GUARD
Catalog No. 5000688098
Encompass
This cut and puncture-resistant sleeve, constructed from SuperFabric material, offers robust protection for industrial applications. Designed with a narrow fit and featuring a gray and black color scheme, it meets stringent ANSI Cut A8 standards for safety.
  • Designed with a narrow size
  • Constructed from SuperFabric material
  • Gray and black color scheme
  • Meets ANSI Cut A8 standards
  • Includes a thumb hole
  • Features snaps at the wrist for a secure fit
  • Sleeve size is 2X-Large
  • Overall length is 14.00 in
  • Provides ANSI Cut Level A5 protection
  • Features 2-Ply construction
  • Offers needlestick protection
  • Comfortable
  • Cut-resistant with ANSI/ISEA Abrasion Resistance Level 6
  • ANSI/ISEA Puncture Resistance Level 2
  • Not flame resistant
Catalog No. 50-006-88098 Supplier MSC Supplier No. 93565232
This cut and puncture-resistant sleeve, constructed from SuperFabric material, offers robust protection for industrial applications. Designed with a narrow fit and featuring a gray and black color scheme, it meets stringent ANSI Cut A8 standards for safety.
  • Designed with a narrow size
  • Constructed from SuperFabric material
  • Gray and black color scheme
  • Meets ANSI Cut A8 standards
  • Includes a thumb hole
  • Features snaps at the wrist for a secure fit
  • Sleeve size is 2X-Large
  • Overall length is 14.00 in
  • Provides ANSI Cut Level A5 protection
  • Features 2-Ply construction
  • Offers needlestick protection
  • Comfortable
  • Cut-resistant with ANSI/ISEA Abrasion Resistance Level 6
  • ANSI/ISEA Puncture Resistance Level 2
  • Not flame resistant