Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neese 1650C Economy Raincoats
SDP

Catalog No. 19412850
Change view
Click to view available options
Black; XXX-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-412-850 Black; XXX-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-412-850 Supplier Neese Supplier No. 10165312BLK3X/DEL/DEL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Size 3X-Large
Material Polyester
Color Black
Closure Type Snap
Coating PVC
Seam Type Sealed
No. of Pockets 2
Size (English) Chest 56 to 58 in.
Size (English) Waist 52 to 54 in.
Size (Metric) Chest 142.2 to 147.3 cm
Length (English) 48 in.
Length (Metric) 121.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.