Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neese Acid Suit 96 Jackets
SDP

Catalog No. 19020316
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Seam Type:
Double-welded
Welded
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
19-020316 Medium Welded
19-020318 X-Large Welded
19-069215 Medium Double-welded
19-046-148A Small Double-welded
19-042902 Large Double-welded
19-069216 X-Large Double-welded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-020316 Supplier Neese Supplier No. 96001001GRNM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Green PVC/polyester knit jackets in a variety of styles

  • Snap front
  • Storm flap
  • Available with attached hood with drawstring or snaps for hood
  • 19-046-148A and 19-042-902 have double-welded seams

Compliance

ASTM D6413: Flame Resistance and ASTM F903: Resistance To Penetration By Liquids

Specifications

Size Medium
Material Polyester, PVC
Color Green
Closure Type Snap
Certifications/Compliance ASTM D6413, ASTM F903
Construction PVC, Polyester Knit
Seam Type Welded
Hood Type Attached
Resistance Flame
Size (English) Chest 40 to 42 in.
Size (Metric) Chest 101.6 to 106.68 cm
For Use With (Application) Chemical, rain, flame
Length (English) 30 in.
Length (Metric) 76.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.