Learn More
Neese Acid Suit 96 Jackets
SDP
Catalog No. 19069215
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Seam Type:
Double-welded
Welded
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
19-069215 Medium Double-welded
19-020316 Medium Welded
19-020318 X-Large Welded
19-046-148A Small Double-welded
19-042902 Large Double-welded
19-069216 X-Large Double-welded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-069215 Supplier Neese Supplier No. 96001011GRNM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Green PVC/polyester knit jackets in a variety of styles
  • Snap front
  • Storm flap
  • Available with attached hood with drawstring or snaps for hood
  • 19-046-148A and 19-042-902 have double-welded seams
Compliance
ASTM D6413: Flame Resistance and ASTM F903: Resistance To Penetration By Liquids
Specifications
Size Medium
Material Polyester, PVC
Color Green
Closure Type Snap
Construction PVC, Polyester Knit
Seam Type Double-welded
Resistance Flame
Length (English) 30 in.
Length (Metric) 76.2 cm