Learn More
Neese Indura 9 oz. FR Lab Coats
SDP
Catalog No. 19412827
Change view
Click to view available options
Color:
Royal Blue
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-412-827 Royal Blue Medium
19-412-831 Royal Blue 3X-Large
19-412-826 Royal Blue Small
19-412-829 Royal Blue X-Large
19-412-830 Royal Blue 2X-Large
19-412-828 Royal Blue Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-412-827 Supplier Neese Supplier No. VI9LCRYM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Flame resistant lab coat offers the standard style coverage as well as FR protection.
  • 9 oz Indura cotton FR fabric - (CAT 2) - 11.5 cal/cm2
  • Classic lab coat styling
  • Leaf collar
  • Snap-up front closure with covered snaps
  • Left breast pocket with pen slot
  • Lower hip side pockets with reinforced bottoms
  • Wrist cuffs with take-up snaps
Specifications
Product Type Flame-resistant Lab Coat
Size Medium
Gender Unisex
Material FR Cotton
Color Royal Blue
Closure Type Snaps
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Standard Length
No. of Pockets 2
Collar Type Leaf
Model VI9LCRY
Arc Flash Rating 11.5 Calories/cm2
Certifications/Compliance ASTM 1506, NFPA 70E
Weight (English) 9 oz.
Weight (Metric) 0.25 kg