Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neese PVC Coveralls
SDP

Catalog No. 19083757
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-083757 2X-Large
19-083754 X-Large
19-069-227 Medium
19-083756 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-083757 Supplier Neese Supplier No. 96001501GRN2X/DEL/DEL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Size 2X-Large
Material PVC, Polyester
Color Green
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper with Snap
Seam Type Welded
Certifications/Compliance ASTM D6413, ASTM F903
For Use With (Application) Sewer works, containment tank cleaning, incidental chemical splash and general industrial applications
Weight (English) 0.81 lb.
Weight (Metric) 0.36 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.