Learn More
Neese Universal 35 Rainwear Bib Trousers, With Safety Fly
SDP
Catalog No. 19043039
Change view
Click to view available options
Color:
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-043039 Yellow 3X-Large
19-020176 - Medium
19-020175 Yellow Large
19-020177 Yellow Small
19-020178 Yellow X-Large
19-020174 Yellow 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-043039 Supplier Neese Supplier No. 35001132YEL3X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provide tough protection
  • Material: PVC / nylon
  • Double stitched and tape sealed seams
  • Flame, abrasion, tear and snag resistant
  • Lightweight
  • Bib front coverage
  • With elastic suspenders
  • Non-conductive snap closures that fastens suspenders onto garment
  • Does not crack or peel in temperatures of -30° to +120°F (-34° to +49°C)
Specifications
Size 3X-Large
Material PVC
Color Yellow
Style Includes elastic suspenders