Learn More
Neese Universal 35 Rainwear Bib Trousers, Without Safety Fly
SDP
Catalog No. 19020172
Change view
Click to view available options
Color:
Yellow
Size:
Small
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-020172 Yellow Small
19-020157 Yellow 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-020172 Supplier Neese Supplier No. 35001121YELS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provide tough protection
  • Material: PVC / nylon
  • Double stitched and tape sealed seams
  • Flame, abrasion, tear and snag resistant
  • Lightweight
  • Bib front coverage
  • With elastic suspenders
  • Non-conductive snap closures that fastens suspenders onto garment
  • Does not crack or peel in temperatures of -30° to +120°F (-34° to +49°C)
Specifications
Size Small
Material Nylon, PVC
Color Yellow
Style Includes elastic suspenders