Learn More
Neese Universal 35 Rainwear Jackets
SDP
Catalog No. 19020165
Change view
Click to view available options
Color:
Green
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-020165 Yellow Medium
19-020164 Yellow Large
19-020166 Yellow Small
19-020167 Yellow X-Large
19-020163 Green Medium
19-020168 Yellow 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-020165 Supplier Neese Supplier No. 35001001YELM
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Material: PVC / nylon
  • Lightweight
  • Flame resistant
  • Minimum length of 30 in. with size adjustments
  • Snap front and storm flap
  • Double stitched and tape sealed seams
  • Attached hood with drawstring closure or stand up collar with snaps for hood
  • Does not crack or peel in temperatures of -30° to +120°F (-34° to +49°C)
Specifications
Size Medium
Material PVC
Color Yellow