Learn More
Neese Universal 35 Rainwear Jackets
SDP
Catalog No. 19020169
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19-020169 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-020169 Supplier Neese Supplier No. 35001002YEL3X
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Material: PVC / nylon
  • Lightweight
  • Flame resistant
  • Minimum length of 30 in. with size adjustments
  • Snap front and storm flap
  • Double stitched and tape sealed seams
  • Attached hood with drawstring closure or stand up collar with snaps for hood
  • Does not crack or peel in temperatures of -30° to +120°F (-34° to +49°C)
Specifications
Size X-Large
Material PVC
Color Yellow