Learn More
Fujifilm Healthcare Americas Corp Hr Series Nefa-Hr (2) Solvent A
Catalog No. NC9517309
Encompass_Preferred
The HR SERIES NEFA-HR (2) SOLVENT A is for use in the HR Series NEFA HR (2) assay. The HR Series NEFA HR (2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum. Package size: 4 x 50 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9517309 Supplier Fujifilm Healthcare Americas Corp Supplier No. 99534791
The HR SERIES NEFA-HR (2) SOLVENT A is for use in the HR Series NEFA HR (2) assay. The HR Series NEFA HR (2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum. Package size: 4 x 50 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.