Learn More
Fujifilm Healthcare Americas Corp Hr Series Nefa-Hr (2) Solvent B
Catalog No. NC9517311
Encompass_Preferred
The HR SERIES NEFA-HR (2) SOLVENT B is for use in the HR Series NEFA HR (2) assay. The HR Series NEFA HR (2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum. Package size: 4 x 25 mL bottles. This product is For In Vitro Diagnostic Use. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9517311 Supplier Fujifilm Healthcare Americas Corp Supplier No. 99335191
The HR SERIES NEFA-HR (2) SOLVENT B is for use in the HR Series NEFA HR (2) assay. The HR Series NEFA HR (2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum. Package size: 4 x 25 mL bottles. This product is For In Vitro Diagnostic Use. Contact supplier for additional information at www.ivd.fujimed.com.