Learn More
Fujifilm Healthcare Americas Corp Nefa Standard Solution
Catalog No. NC9517312
Encompass_Preferred
The NEFA Standard Solution is designed to be used to establish points of reference in the measurement of free fatty acids in serum or plasma when used with the HR NEFA HR (2) series of reagents. The NEFA standard solution contains Oleic Acid and a Stabilizer that has a concentration of 1.0 mEq/L. Package size: 4 x10 mL bottles. This product is for In Vitro Diagnostic Use. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9517312 Supplier Fujifilm Healthcare Americas Corp Supplier No. 27676491
The NEFA Standard Solution is designed to be used to establish points of reference in the measurement of free fatty acids in serum or plasma when used with the HR NEFA HR (2) series of reagents. The NEFA standard solution contains Oleic Acid and a Stabilizer that has a concentration of 1.0 mEq/L. Package size: 4 x10 mL bottles. This product is for In Vitro Diagnostic Use. Contact supplier for additional information at www.ivd.fujimed.com.