Learn More
Selleck Chemical LLC Nefazodone hydrochloride 10mg
SDP
Supplier:  Selleck Chemical LLC S539810MG
Encompass_Preferred
Nefazodone hydrochloride (BMY-13754-1) is the hydrochloride salt form of nefazodone, which is an atypical antidepressant showing antagonistic activity on serotonin reuptake. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3161