Learn More
Cayman Chemical Nefazodone (hydrochloride), Purity: ≥98%, CAS: 82752-99-6, 1 g
SDP
Catalog No. 502889448
Encompass_Preferred
A serotonin, norepinephrine, and dopamine reuptake inhibitor (Kis = 137, 570, 2,380 nM, respectively); an antagonist of 5-HT2A, 5-HT1A, and histamine H1 receptors, as well as α1- and α2-ARs (Kis = 7.1, 52, 30, 5.5, and 84 nM, respectively); reduces immobility time in the tail suspension test in gerbils at 100 mg/kg; toxic to isolated human hepatocytes in a cultured sandwich hepatocyte preparation at 30 µM
Catalog No. 50-288-9448 Supplier Cayman Chemical Supplier No. 100126421 G
A serotonin, norepinephrine, and dopamine reuptake inhibitor (Kis = 137, 570, 2,380 nM, respectively); an antagonist of 5-HT2A, 5-HT1A, and histamine H1 receptors, as well as α1- and α2-ARs (Kis = 7.1, 52, 30, 5.5, and 84 nM, respectively); reduces immobility time in the tail suspension test in gerbils at 100 mg/kg; toxic to isolated human hepatocytes in a cultured sandwich hepatocyte preparation at 30 µM