Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4747MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IF ,Flow, IHC. Conjugate/Tag: Unconjugated. Clone: NFL/736. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Brain, Neuroblastoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human NEFL protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 68kDa.Chromosome: 8p21.2. Gene Symbol: NEFL. Gene ID: 4747. Ensemble: ENSG00000277586. Uniprot: NP_006149.
Catalog No. 50-188-5013