Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4744MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IHC. Conjugate/Tag: Unconjugated. Clone: SPM203. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Brain, Neuroblastoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Triton-X 100 insoluble protein fraction of rat brain. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 200kDa.Chromosome: 22q12.2. Gene Symbol: NEFH. Gene ID: 4744. Ensemble: ENSG00000100285. Uniprot: NP_066554.
Catalog No. 50-188-5009