Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM2X1201P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IHC. Conjugate/Tag: Unconjugated. Clone: SPM145. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Cerebellum, Neuroblastoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Semi-purified human neurofilaments from spinal cord. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ent light polypeptide, Neurofilament protein, light chain, Neurofilament subunit NF-L, Neurofilament triplet L protein, NF-L, NF68, anti-NEFH antibody, anti-Neurofilament H antibody, anti-Neurofilament Heavy Poly.Chromosome: 22q12.2 (NEFH) & 8p21.2 (NEFL). Gene Symbol: NEFH & NEFL. Gene ID: 4744. Ensemble: ENSG00000100285. Uniprot: NP_066554.
Catalog No. 50-188-5461