Learn More
Apexbio Technology LLC Nefiracetam 77191-36-7 250mg
SDP
Catalog No. 5000087680
Encompass_Preferred
Nefiracetam (CAS 77191-36-7) is a small molecule modulator that enhances neuronal signaling across GABAergic cholinergic and monoaminergic pathways It has been shown to potentiate synaptic transmission and is investigated for its ability to counteract convulsions induced by Ro 5-4864 a peripheral benzodiazepine receptor agonist Nefiracetam s neuropharmacological activity positions it as a valuable tool in research on cognitive enhancement synaptic plasticity and seizure modulation and it is currently under evaluation in Phase 2 clinical studies
Catalog No. 50-000-87680 Supplier Apexbio Technology LLC Supplier No. B1532250
Nefiracetam (CAS 77191-36-7) is a small molecule modulator that enhances neuronal signaling across GABAergic cholinergic and monoaminergic pathways It has been shown to potentiate synaptic transmission and is investigated for its ability to counteract convulsions induced by Ro 5-4864 a peripheral benzodiazepine receptor agonist Nefiracetam s neuropharmacological activity positions it as a valuable tool in research on cognitive enhancement synaptic plasticity and seizure modulation and it is currently under evaluation in Phase 2 clinical studies